Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adamts17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033877.4 | NP_001029049.2 | 1 - 2 | 177 | 142 | |
GenBank mRNA | AK045148.1 | - | 1 - 2 | 178 | 142 | |
AK053131.1 | - | 1 - 2 | 178 | 142 | ||
BC141170.1 | - | 1 - 2 | 129 | 142 |
Set Membership: |
Amplicon length greater than or equal to 101 Protease Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons |