Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bin3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021328.3 | NP_067303.1 | 2 - 3 | 108 | 67 | |
GenBank mRNA | AF244361.1 | - | 2 - 3 | 88 | 67 | |
AF271733.1 | - | 2 - 3 | 151 | 67 | ||
AK005992.1 | - | 2 - 3 | 117 | 67 | ||
AK040685.1 | - | 2 - 3 | 108 | 67 | ||
AK170915.1 | - | 2 - 3 | 149 | 67 | ||
AK210433.1 | - | 2 - 3 | 89 | 67 | ||
BC026543.1 | - | 2 - 3 | 93 | 5040094 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |