Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0300023064.mm.1 |
NM_027808.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300023064.mm.1 |
NM_027808.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300023064.mm.1 |
TC0300002912.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300023066.mm.1 |
NM_027808.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300023066.mm.1 |
NM_027808.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300023066.mm.1 |
TC0300002912.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300023067.mm.1 |
NM_027808.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300023067.mm.1 |
NM_027808.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300023067.mm.1 |
TC0300002912.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300023068.mm.1 |
NM_027808.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300023068.mm.1 |
NM_027808.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300023068.mm.1 |
TC0300002912.mm.1 |
MTA TC |