Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpatch2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026367.4 | NP_080643.4 | 7 - 8 | 1318 | 76 | |
GenBank mRNA | AK029990.1 | - | 7 - 8 | 1302 | 76 | |
AK030026.1 | - | 7 - 8 | 1317 | 76 | ||
AK083471.1 | - | 6 - 7 | 475 | 76 | ||
BC054810.1 | - | 7 - 8 | 1290 | 6839419 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |