Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smyd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027188.3 | NP_081464.1 | 6 - 7 | 650 | 127 | |
GenBank mRNA | AK010447.1 | - | 6 - 7 | 684 | 127 | |
AK043988.1 | - | 6 - 7 | 660 | 127 | ||
AK163057.1 | - | 6 - 7 | 667 | 127 | ||
AK167875.1 | - | 6 - 7 | 665 | 127 | ||
BC019663.1 | - | 6 - 7 | 652 | 127 | ||
BC038482.1 | - | 6 - 7 | 531 | 127 | ||
BC052431.1 | - | 6 - 7 | 650 | 6837012 | 127 | |
BC061485.1 | - | 6 - 7 | 633 | 5370924 | 127 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |