Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Opn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010098.3 | NP_034228.1 | 2 - 3 | 787 | 133 | |
GenBank mRNA | AF140241.1 | - | 2 - 3 | 745 | 133 | |
AK043135.1 | - | 2 - 3 | 788 | 133 | ||
AK132822.1 | - | 2 - 3 | 724 | 133 | ||
BC104486.1 | - | 3 - 4 | 637 | 133 | ||
BC104487.1 | - | 3 - 4 | 660 | 132 | ||
BC105646.1 | - | 3 - 4 | 584 | 133 | ||
BC105919.1 | - | 3 - 4 | 584 | 133 | ||
BC139805.1 | - | 3 - 4 | 585 | 133 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor Mammalian Gene Collection Mus musculus Probe spans exons |