Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr64 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029453.2 | NP_083729.2 | 26 - 27 | 3273 | 114 | |
GenBank mRNA | AK015753.1 | - | 14 - 15 | 1620 | 114 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |