Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxo28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175127.2 | NP_780336.1 | 2 - 3 | 388 | 78 | |
GenBank mRNA | AK076211.1 | - | 2 - 3 | 394 | 78 | |
AK077653.1 | - | 2 - 3 | 401 | 78 | ||
AK122295.1 | - | 2 - 3 | 373 | 78 | ||
AK162091.1 | - | 2 - 3 | 357 | 78 | ||
AK162188.1 | - | 2 - 3 | 269 | 78 | ||
BC055458.1 | - | 1 - 2 | 90 | 78 | ||
BC094542.1 | - | 2 - 3 | 388 | 6408744 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |