Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Capn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009794.3 | NP_033924.2 | 7 - 8 | 1026 | 122 | |
GenBank mRNA | AF015038.1 | - | 7 - 8 | 972 | 122 | |
AK052141.1 | - | 7 - 8 | 1001 | 122 | ||
AK053832.1 | - | 7 - 8 | 2312 | 122 | ||
AK078787.1 | - | 3 - 4 | 1655 | 122 | ||
AK150695.1 | - | 7 - 8 | 1000 | 122 | ||
AK152094.1 | - | 7 - 8 | 1000 | 122 | ||
AK152790.1 | - | 7 - 8 | 1002 | 122 | ||
AK152994.1 | - | 7 - 8 | 1002 | 122 | ||
AK154855.1 | - | 7 - 8 | 1040 | 122 | ||
BC054726.1 | - | 7 - 8 | 1026 | 5703563 | 122 | |
D38117.1 | - | 7 - 8 | 1067 | 122 | ||
Y10139.1 | - | 7 - 8 | 908 | 122 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protease Mammalian Gene Collection Mus musculus Probe spans exons |