Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Alg14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024178.2 | NP_077140.1 | 2 - 3 | 325 | 65 | |
GenBank mRNA | AK011733.1 | - | 2 - 3 | 371 | 65 | |
BC002278.1 | - | 2 - 3 | 325 | 3496897 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |