Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp51 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_020010.2 | NP_064394.2 | 4 - 5 | 962 | 114 | |
GenBank mRNA | AF166266.1 | - | 4 - 5 | 714 | 114 | |
AK028355.1 | - | 4 - 5 | 655 | 114 | ||
AK028815.1 | - | 4 - 5 | 719 | 114 | ||
AK031059.1 | - | 4 - 5 | 695 | 114 | ||
AK075764.1 | - | 4 - 5 | 695 | 114 | ||
AK076983.1 | - | 4 - 5 | 695 | 114 | ||
BC031813.1 | - | 4 - 5 | 689 | 4193738 | 114 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |