Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adamdec1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021475.2 | NP_067450.1 | 10 - 11 | 1027 | 83 | |
GenBank mRNA | AJ242912.1 | - | 10 - 11 | 1027 | 83 | |
AK156586.1 | - | 10 - 11 | 1042 | 83 | ||
BC026542.1 | - | 10 - 11 | 1050 | 84 | ||
BC046324.1 | - | 10 - 11 | 1018 | 6394673 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protease Mammalian Gene Collection Mus musculus Probe spans exons |