Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdc42bpa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033285.1 | NP_001028457.1 | 24 - 25 | 3146 | 107 | |
GenBank mRNA | BC158017.1 | - | 24 - 25 | 3146 | 9007323 | 107 |
BC158095.1 | - | 23 - 24 | 3107 | 9007382 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |