Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gmfb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022023.2 | NP_071306.2 | 1 - 2 | 144 | 115 | |
GenBank mRNA | AB050013.1 | - | 1 - 2 | 86 | 115 | |
AK013947.1 | - | 1 - 2 | 101 | 115 | ||
AK142014.1 | - | 1 - 2 | 116 | 115 | ||
BC040233.1 | - | 1 - 2 | 78 | 5364203 | 115 | |
BC049134.1 | - | 1 - 2 | 82 | 6831501 | 115 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |