Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010551.3 | NP_034681.2 | 12 - 13 | 2095 | 78 | |
GenBank mRNA | AF175292.1 | - | 12 - 13 | 2095 | 78 | |
AK220455.1 | - | 12 - 13 | 2105 | 78 | ||
BC058709.1 | - | 12 - 13 | 2128 | 6841305 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |