Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Naa16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025832.2 | NP_080108.1 | 15 - 16 | 2005 | 159 | |
GenBank mRNA | AK005056.1 | - | 15 - 16 | 2015 | 159 | |
AK028432.1 | - | 15 - 16 | 2116 | 159 | ||
AK152498.1 | - | 15 - 16 | 2015 | 159 | ||
BC048727.1 | - | 3 - 4 | 546 | 159 | ||
BC052445.1 | - | 15 - 16 | 2005 | 6412745 | 159 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |