Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abca4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007378.1 | NP_031404.1 | 12 - 13 | 1850 | 67 | |
GenBank mRNA | AF000149.1 | - | 12 - 13 | 1850 | 67 | |
BC057853.1 | - | 12 - 13 | 1850 | 6492139 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |