Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lamc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011836.3 | NP_035966.2 | 6 - 7 | 1369 | 61 | |
GenBank mRNA | AF079520.1 | - | 6 - 7 | 1359 | 61 | |
AF083372.1 | - | 6 - 7 | 1306 | 61 | ||
AK132948.1 | - | 3 - 4 | 354 | 61 | ||
BC096366.1 | - | 6 - 7 | 1369 | 2865309 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |