Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cpxm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019696.2 | NP_062670.2 | 3 - 4 | 541 | 88 | |
GenBank mRNA | AF077738.1 | - | 3 - 4 | 486 | 88 | |
BC003713.1 | - | 3 - 4 | 474 | 3593211 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protease Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |