Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gabra5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_176942.4 | NP_795916.1 | 1 - 2 | 275 | 147 | |
GenBank mRNA | AK038476.1 | - | 1 - 2 | 277 | 148 | |
BC062112.1 | - | 1 - 2 | 273 | 6815819 | 147 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Ion Channel Mammalian Gene Collection Mus musculus Probe spans exons |