Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ctc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013256.2 | NP_001013274.2 | 5 - 6 | 1010 | 128 | |
NM_001143790.1 | NP_001137262.1 | 5 - 6 | 1010 | 128 | ||
NM_001281464.1 | NP_001268393.1 | 5 - 6 | 881 | 128 | ||
NM_001281465.1 | NP_001268394.1 | 5 - 6 | 994 | 128 | ||
GenBank mRNA | AK038651.1 | - | 5 - 6 | 797 | 128 | |
AK044055.1 | - | 5 - 6 | 789 | 128 | ||
AK145919.1 | - | 5 - 6 | 825 | 128 | ||
AK165906.1 | - | 5 - 6 | 810 | 128 | ||
AK217260.1 | - | 2 - 3 | 363 | 128 | ||
BC066078.1 | - | 5 - 6 | 825 | 128 | ||
BC072625.1 | - | 5 - 6 | 838 | 128 | ||
BC147495.1 | - | 5 - 6 | 808 | 9056325 | 128 | |
BC147497.1 | - | 5 - 6 | 808 | 9056354 | 128 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |