Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc25a35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028048.2 | NP_082324.1 | 2 - 3 | 971 | 73 | |
GenBank mRNA | AK007453.1 | - | 2 - 3 | 973 | 73 | |
AK171131.1 | - | 2 - 3 | 978 | 73 | ||
BC004569.1 | - | 2 - 3 | 411 | 73 | ||
BC019996.1 | - | 2 - 3 | 913 | 4207271 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |