Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Specc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029936.3 | NP_001025107.1 | 6 - 7 | 2402 | 92 | |
NM_001281818.1 | NP_001268747.1 | 8 - 9 | 2619 | 92 | ||
GenBank mRNA | AK220465.1 | - | 6 - 7 | 2329 | 92 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mus musculus Probe spans exons |