Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gucy2f | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007576.2 | NP_001007577.1 | 2 - 3 | 972 | 75 | |
GenBank mRNA | AK044234.1 | - | 2 - 3 | 973 | 75 | |
AY651761.1 | - | 2 - 3 | 967 | 75 | ||
BC115714.1 | - | 1 - 2 | 802 | 40085764 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |