Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_181820.2 | NP_861541.2 | 4 - 5 | 711 | 97 | |
GenBank mRNA | AK050105.1 | - | 3 - 4 | 606 | 97 | |
AY236493.1 | - | 4 - 5 | 674 | 97 | ||
AY263162.1 | - | 4 - 5 | 682 | 97 | ||
BC040466.1 | - | 4 - 5 | 623 | 97 | ||
BC055939.1 | - | 3 - 4 | 440 | 97 | ||
BC106105.1 | - | 4 - 5 | 682 | 97 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |