Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Leng8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172736.3 | NP_766324.1 | 13 - 14 | 2060 | 60 | |
GenBank mRNA | AK034310.1 | - | 13 - 14 | 2065 | 60 | |
AK170629.1 | - | 13 - 14 | 2068 | 60 | ||
BC034448.1 | - | 5 - 6 | 784 | 60 | ||
BC041775.1 | - | 12 - 13 | 2565 | 60 | ||
BC042658.1 | - | 13 - 14 | 2048 | 5365041 | 60 | |
BC066768.1 | - | 13 - 14 | 2051 | 6407146 | 60 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |