Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ston2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175367.6 | NP_780576.1 | 3 - 4 | 861 | 55 | |
GenBank mRNA | AK036612.1 | - | 3 - 4 | 862 | 55 | |
BC132531.1 | - | 2 - 3 | 447 | 40130808 | 55 | |
BC138054.1 | - | 2 - 3 | 447 | 8861073 | 55 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |