Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scrn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027268.2 | NP_081544.1 | 2 - 3 | 300 | 61 | |
GenBank mRNA | AK012765.1 | - | 2 - 3 | 328 | 61 | |
AK032573.1 | - | 2 - 3 | 327 | 61 | ||
AK050481.1 | - | 2 - 3 | 318 | 61 | ||
AK082953.1 | - | 2 - 3 | 300 | 61 | ||
AK089968.1 | - | 2 - 3 | 290 | 61 | ||
AK129084.1 | - | 2 - 3 | 287 | 61 | ||
AK145904.1 | - | 2 - 3 | 302 | 61 | ||
BC023889.1 | - | 2 - 3 | 287 | 5252046 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |