Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rif1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175238.5 | NP_780447.4 | 22 - 23 | 2659 | 118 | |
GenBank mRNA | AK088688.1 | - | 10 - 11 | 2351 | 118 | |
AY428571.2 | - | 22 - 23 | 2643 | 118 | ||
AY585206.1 | - | 21 - 22 | 2652 | 118 | ||
JX255737.1 | - | 21 - 22 | 2595 | 118 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |