Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pde11a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081033.1 | NP_001074502.1 | 17 - 18 | 2535 | 76 | |
GenBank mRNA | BC158125.1 | - | 17 - 18 | 2675 | 9088197 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |