Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ubr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK050359.1 | - | 1 - 2 | 120 | 80 | |
BC157930.1 | - | 9 - 10 | 1551 | 80 | ||
BC172022.1 | - | 9 - 10 | 1552 | 80 | ||
BC172146.1 | - | 29 - 30 | 4118 | 80 | ||
DQ924536.1 | - | 29 - 30 | 4415 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |