Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnh7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133207.2 | NP_573470.2 | 11 - 12 | 2820 | 153 | |
GenBank mRNA | AJ291608.1 | - | 11 - 12 | 2627 | 153 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Ion Channel Mus musculus Probe spans exons |