Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rassf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175445.4 | NP_780654.1 | 7 - 8 | 913 | 68 | |
GenBank mRNA | AK028384.1 | - | 7 - 8 | 817 | 68 | |
AK172906.1 | - | 7 - 8 | 719 | 68 | ||
BC057402.1 | - | 7 - 8 | 735 | 5698333 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |