Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ubr7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025666.2 | NP_079942.1 | 1 - 2 | 205 | 64 | |
GenBank mRNA | AK011968.1 | - | 1 - 2 | 76 | 64 | |
AK017542.1 | - | 1 - 2 | 160 | 64 | ||
AK088208.1 | - | 1 - 2 | 207 | 64 | ||
AK146308.1 | - | 1 - 2 | 206 | 64 | ||
AK149989.1 | - | 1 - 2 | 160 | 64 | ||
AK155611.1 | - | 1 - 2 | 76 | 64 | ||
BC094323.1 | - | 1 - 2 | 213 | 64 | ||
BC094423.1 | - | 1 - 2 | 163 | 6812389 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |