Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dpm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010073.2 | NP_034203.1 | 1 - 2 | 57 | 62 | |
GenBank mRNA | AB013360.1 | - | 1 - 2 | 29 | 62 | |
AK002468.1 | - | 1 - 2 | 41 | 62 | ||
AK004241.1 | - | 1 - 2 | 32 | 62 | ||
BC008256.1 | - | 1 - 2 | 57 | 2648752 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |