Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sdccag3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026563.3 | NP_080839.2 | 2 - 3 | 312 | 101 | |
GenBank mRNA | AK014885.1 | - | 2 - 3 | 281 | 101 | |
AK021273.1 | - | 2 - 3 | 303 | 101 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Mus musculus Probe spans exons |