Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cytip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_139200.4 | NP_631939.1 | 5 - 6 | 541 | 87 | |
GenBank mRNA | AF192525.1 | - | 5 - 6 | 461 | 87 | |
AK152784.1 | - | 5 - 6 | 523 | 87 | ||
AK157859.1 | - | 5 - 6 | 542 | 87 | ||
AK166254.1 | - | 5 - 6 | 542 | 87 | ||
AK169575.1 | - | 5 - 6 | 527 | 87 | ||
AK170544.1 | - | 5 - 6 | 542 | 87 | ||
AK170796.1 | - | 5 - 6 | 542 | 87 | ||
AK171467.1 | - | 5 - 6 | 542 | 87 | ||
AK172071.1 | - | 5 - 6 | 598 | 87 | ||
AK172380.1 | - | 5 - 6 | 542 | 87 | ||
BC007144.1 | - | 5 - 6 | 530 | 3487179 | 87 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |