Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Asb6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133346.2 | NP_579924.1 | 3 - 4 | 511 | 106 | |
GenBank mRNA | AF398967.1 | - | 2549 | 106 | ||
AK010909.1 | - | 3 - 4 | 492 | 106 | ||
AK049312.1 | - | 3 - 4 | 500 | 106 | ||
AK088890.1 | - | 3 - 4 | 446 | 106 | ||
AY665653.1 | - | 3 - 4 | 397 | 106 | ||
BC019192.1 | - | 3 - 4 | 503 | 5035395 | 106 | |
BC085485.1 | - | 3 - 4 | 468 | 106 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |