Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adarb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001289530.1 | NP_001276459.1 | 6 - 7 | 1889 | 79 | |
NM_052977.5 | NP_443209.2 | 6 - 7 | 1889 | 79 | ||
NR_110345.1 | 6 - 7 | 1889 | 79 | |||
GenBank mRNA | AF232942.1 | - | 6 - 7 | 1506 | 79 | |
AF270495.1 | - | 6 - 7 | 1606 | 79 | ||
AK033916.1 | - | 6 - 7 | 1828 | 79 | ||
AK045538.1 | - | 6 - 7 | 1828 | 79 | ||
AK046083.1 | - | 6 - 7 | 1833 | 79 | ||
AK134964.1 | - | 6 - 7 | 1891 | 79 | ||
AK139079.1 | - | 6 - 7 | 1847 | 79 | ||
AK139273.1 | - | 6 - 7 | 1847 | 79 | ||
BC052426.1 | - | 6 - 7 | 1814 | 6835942 | 79 | |
BC059822.1 | - | 6 - 7 | 1842 | 6844585 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |