Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fcnb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010190.1 | NP_034320.1 | 3 - 4 | 250 | 99 | |
GenBank mRNA | AF063217.1 | - | 3 - 4 | 223 | 99 | |
AK010913.1 | - | 2 - 3 | 248 | 99 | ||
AK151676.1 | - | 3 - 4 | 258 | 99 | ||
BC107220.1 | - | 3 - 4 | 250 | 40051808 | 99 | |
BC107221.1 | - | 3 - 4 | 250 | 40051810 | 99 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |