Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gins1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027014.1 | NP_081290.1 | 6 - 7 | 644 | 74 | |
GenBank mRNA | AK013116.1 | - | 6 - 7 | 637 | 74 | |
AK129081.1 | - | 5 - 6 | 435 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mus musculus 1700 Probe spans exons |