Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Strbp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009261.3 | NP_033287.2 | 15 - 16 | 2059 | 90 | |
GenBank mRNA | AK006314.1 | - | 2 - 3 | 242 | 90 | |
AK078956.1 | - | 6 - 7 | 850 | 90 | ||
BC014710.1 | - | 16 - 17 | 2143 | 4502840 | 90 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |