Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0200029051.mm.1 |
NM_001083125.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200029051.mm.1 |
NM_001083125.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200029051.mm.1 |
NM_001083126.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200029051.mm.1 |
NM_001083126.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200029051.mm.1 |
NM_001083127.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200029051.mm.1 |
NM_001083127.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200029051.mm.1 |
NM_008500.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200029051.mm.1 |
NM_008500.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200029051.mm.1 |
TC0200003378.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200029049.mm.1 |
NM_001083125.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200029049.mm.1 |
NM_001083125.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200029049.mm.1 |
NM_001083126.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200029049.mm.1 |
NM_001083126.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200029049.mm.1 |
NM_001083127.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200029049.mm.1 |
NM_001083127.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200029049.mm.1 |
NM_008500.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200029049.mm.1 |
NM_008500.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200029049.mm.1 |
TC0200003378.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200029050.mm.1 |
NM_001083125.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200029050.mm.1 |
NM_001083125.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200029050.mm.1 |
NM_001083126.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200029050.mm.1 |
NM_001083126.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200029050.mm.1 |
NM_001083127.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200029050.mm.1 |
NM_001083127.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200029050.mm.1 |
NM_008500.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200029050.mm.1 |
NM_008500.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200029050.mm.1 |
TC0200003378.mm.1 |
MTA TC |