Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nxt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001110159.1 | NP_001103629.1 | 1 - 2 | 194 | 132 | |
GenBank mRNA | BY091455.1 | - | 1 - 2 | 178 | 132 |
Set Membership: |
Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |