Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0200040482.mm.1 |
NM_175225.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200040482.mm.1 |
NR_110350.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200040482.mm.1 |
TC0200004725.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200040481.mm.1 |
NM_001159640.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200040481.mm.1 |
NM_001159641.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200040481.mm.1 |
NM_001289611.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200040481.mm.1 |
NM_175225.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200040481.mm.1 |
NR_110350.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200040481.mm.1 |
NR_110351.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200040481.mm.1 |
NR_110352.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200040481.mm.1 |
TC0200004725.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200040477.mm.1 |
NM_001159640.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200040477.mm.1 |
NM_001159641.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200040477.mm.1 |
NM_001289611.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200040477.mm.1 |
NM_175225.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200040477.mm.1 |
NR_110350.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200040477.mm.1 |
NR_110351.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200040477.mm.1 |
NR_110352.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200040477.mm.1 |
TC0200004725.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200040484.mm.1 |
NM_001159640.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200040484.mm.1 |
NM_001159641.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200040484.mm.1 |
NM_001289611.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200040484.mm.1 |
NM_175225.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200040484.mm.1 |
NR_110350.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200040484.mm.1 |
NR_110351.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200040484.mm.1 |
NR_110352.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200040484.mm.1 |
TC0200004725.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200040480.mm.1 |
NM_001159640.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200040480.mm.1 |
NM_001159641.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200040480.mm.1 |
NM_001289611.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200040480.mm.1 |
NM_175225.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200040480.mm.1 |
NR_110350.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200040480.mm.1 |
NR_110351.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200040480.mm.1 |
NR_110352.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200040480.mm.1 |
TC0200004725.mm.1 |
MTA TC |