Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scrn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029022.1 | NP_083298.1 | 4 - 5 | 601 | 145 | |
GenBank mRNA | AK014701.1 | - | 4 - 5 | 595 | 145 | |
AK028703.1 | - | 4 - 5 | 575 | 145 | ||
AK143725.1 | - | 4 - 5 | 598 | 145 | ||
AK165936.1 | - | 4 - 5 | 598 | 145 | ||
BC015296.1 | - | 3 - 4 | 535 | 145 | ||
BC139390.1 | - | 4 - 5 | 596 | 8862412 | 145 | |
BC139391.1 | - | 4 - 5 | 596 | 8862413 | 145 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |