Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tekt3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027660.1 | NP_081936.1 | 2 - 3 | 726 | 105 | |
GenBank mRNA | AY252101.1 | - | 2 - 3 | 726 | 105 | |
BC127605.1 | - | 2 - 3 | 624 | 40057157 | 105 | |
BC128270.1 | - | 2 - 3 | 624 | 40057161 | 105 | |
BC139074.1 | - | 2 - 3 | 693 | 8862096 | 105 | |
BC139075.1 | - | 2 - 3 | 693 | 8862097 | 105 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |