Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1100006482.mm.1 |
NM_175002.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100006482.mm.1 |
NM_175002.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100006482.mm.1 |
TC1100000808.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100006503.mm.1 |
NM_175002.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100006503.mm.1 |
NM_175002.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100006503.mm.1 |
TC1100000808.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100006504.mm.1 |
NM_175002.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100006504.mm.1 |
NM_175002.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100006504.mm.1 |
TC1100000808.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100006510.mm.1 |
NM_175002.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100006510.mm.1 |
NM_175002.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100006510.mm.1 |
TC1100000808.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100006501.mm.1 |
NM_175002.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100006501.mm.1 |
NM_175002.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100006501.mm.1 |
TC1100000808.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100006505.mm.1 |
NM_175002.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100006505.mm.1 |
NM_175002.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100006505.mm.1 |
TC1100000808.mm.1 |
MTA TC |