Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Accs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290782.1 | NP_001277711.1 | 11 - 12 | 1152 | 60 | |
NM_183220.3 | NP_899043.1 | 11 - 12 | 1207 | 60 | ||
GenBank mRNA | AK040509.1 | - | 8 - 9 | 2952 | 60 | |
BC039569.1 | - | 11 - 12 | 1184 | 1449080 | 60 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |