Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Catsperb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK040480.1 | - | 6 - 7 | 861 | 129 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
kinesin family member 4, pseudogene |
Kif4-ps |
4930463M05Rik, Kif4b |
-400.1kb | Sense | Chr. 12 - 101145609 - 101149273 on NCBI Build GRCm38 |
Downstream |
tandem C2 domains, nuclear |
Tc2n |
4933406D09Rik, Mtac2d1, Tac2-N |
94.5kb | Anti-sense | Chr. 12 - 101645446 - 101719000 on NCBI Build GRCm38 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Mus musculus Probe spans exons |